Osmium(VIII) oxide, 4% aq. solution, Alfa Aesar™

Manufacturer: Alfa Aesar 453859W

Quantity 2mL

 View more versions of this product

Catalog No. AA453859W

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20816-12-0
    Physical Form Liquid
    Molecular Formula OsO4
    Formula Weight 254.1
    MDL Number MFCD00011150
    Chemical Name or Material Osmium(VIII) oxide

    Specifications

    Formula Weight 254.1
    Quantity 2mL
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 244-058-7
    Physical Form Liquid
    Molecular Formula OsO4
    MDL Number MFCD00011150
    CAS 20816-12-0
    Vapor Pressure 17mm Hg at 20°C
    Health Hazard 1 H301-H310-H332-H315-H318: Toxic if swallowed. Fatal in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye damage.
    Chemical Name or Material Osmium(VIII) oxide

    H301-H310-H332-H315-H318: Toxic if swallowed. Fatal in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit