Osmium(VIII) oxide, 99.8% (Metals basis), Os 74.4% min., Alfa Aesar™

Manufacturer: Alfa Aesar 1210303

Quantity 1g

 View more versions of this product

Catalog No. AA1210303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20816-12-0
    Physical Form Crystalline
    Assay Percent Range 99.8% (metals basis)
    Molecular Formula OsO4
    Formula Weight 254.1
    MDL Number MFCD00011150
    Synonym Osmic acid
    Chemical Name or Material Osmium(VIII) oxide

    Specifications

    Formula Weight 254.1
    Quantity 1g
    DOT Information DOT Class: 6.1, Packaging Group: I
    Health Hazard 2 Keep container tightly closed and in a well-ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    EINECS Number 244-058-7
    Physical Form Crystalline
    Assay Percent Range 99.8% (metals basis)
    Molecular Formula OsO4
    Appearance Crystalline
    MDL Number MFCD00011150
    Odor Like chlorine
    CAS 20816-12-0
    Vapor Pressure 10 hPa (8mm Hg) at 20°C
    Synonym Osmic acid
    Health Hazard 1 H300-H310-H330-H314: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage.
    Chemical Name or Material Osmium(VIII) oxide

    Keep container tightly closed and in a well-ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H300-H310-H330-H314: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit