Learn More
Osmosis Simulation Activity Model
SCI_ED
Focuses on osmosis.
Manufacturer:  Neo/Sci Corporation 301125
Catalog No. S66659
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This striking demonstration of osmosis will show water diffusion across a semipermeable membrane, from an area of higher concentration to an area of lower concentration.
  • Results can be quantified by measuring the amount of liquid that travels from one side to the other
  • Repeatable with many different solutes in varying concentrations
  • Includes a display stand
  • Dimensions: 1.5 × 6.5 × 7 in. (3.8 × 16.5 × 17.8cm)
Specifications
Specifications
Provides visual demonstration of osmosis
Display stand
Osmosis Simulation Activity Model