Sekisui Diagnostics OSOM™ Ultra Flu A and B Rapid Test

OSOM Ultra Flu A&B is a rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens

Manufacturer: Sekisui Diagnostics 1009

 View more versions of this product

Catalog No. 22-321-108

  Check Availability
Add to Cart

Includes: 25 Swabs only

    Specifications

    Certifications/Compliance Cleared for use with multiple viral transport media (VTM) types
    Description OSOM Ultra Flu A and B Sample Swabs
    CPT Code Flu A: 87804; Flu B: 87804-59 (modifier used to indicate separate test)
    Detectable Analytes Influenza Type A and Type B
    For Use With (Application) For the detection of Influenza type A and type B nucleoprotein antigens
    Includes 25 sample swabs only
    Quantity 25/Pk.
    Sample Type Nasal swab, NP swab as well as nasal aspirate/wash
    Sensitivity Influenza A: >>90%; Influenza B: >80% vs culture
    Storage Requirements Store at room temperature.
    Test Time 10 to 15 minutes
    Product Type OSOM Ultra Flu A and B Sample Swabs

    Recommended Storage

    Store at room temperature.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit