Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sekisui Diagnostics OSOM™ Ultra Flu A&B Test Control Swab Kit

OSOM Ultra Flu A&B is a rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens.

Manufacturer:  Sekisui Diagnostics 1005

Catalog No. 22-321-114


Add to cart

Includes: 5 x positive Flu A/B swabs and 5 x Negative Flu A/B swabs

Description

Description

Improved Outcomes: Rapid and accurate diagnosis of Influenza enables a choice of antiviral therapy, helps avoid the overuse of antibiotics, and prevents healthcare costs related to unnecessary testing and treatment.

Patient Experience: Streamlines lab's workflow while reducing patient wait time and unnecessary use of antibiotics.

  • For the detection of Influenza type A and type B nucleoprotein antigens
  • Able to meet FDA reclassification standards
  • Multiple specimen types: nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens
  • Cleared for use with multiple viral transport media (VTM) types
  • Accuracy: Influenza A Sensitivity: >90%; Influenza B: >85% vs culture (across multiple sample types)
  • Fast: Obtain positive results in 10 to 15 minutes
  • Flexible: Runs from nasal swab, NP swab as well as nasal aspirate/wash
  • Workflow: approved for use with multiple viral transport media types
  • QC Inside: 2 extra no charge cassettes to the kit

OSOM Ultra Flu A and B Control Kit (Cat. No. 22321114): 5 x positive Flu A/B swabs and 5 x Negative Flu A/B swabs
OSOM Ultra Flu A and B Nasopharyngeal Wash/Aspirate Kit (Cat. No. 22321106): 50 pipets and 5mL extraction reagent
OSOM Ultra Flu A and B Swab Transport Kit (Cat. No. 22321107): 25 swabs in transport tubes

Recommended Storage

Store at room temperature.

Specifications

Specifications

For the detection of Influenza type A and type B nucleoprotein antigens
10 to 15 minutes
Cleared for use with multiple viral transport media (VTM) types
Flu A: 87804; Flu B: 87804-59 (modifier used to indicate separate test)
Influenza Type A and Type B
OSOM Ultra Flu A&B Test Control Swab Kit
Influenza A: >>90%; Influenza B: >80% vs culture
Nasal Aspirate/Wash, Nasal Swab, Nasopharyngeal Swab
Room Temperature
OSOM Ultra Flu A and B Control Kit
5 x positive Flu A/B swabs and 5 x Negative Flu A/B swabs
25/Pk.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit