Learn More
Osteoactivin/GPNMB Mouse anti-Human, DyLight 755, Clone: 7C10E5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237351IR
Catalog No. NBP237351IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Osteoactivin/GPNMB
7C10E5
Western Blot, ELISA, Immunohistochemistry
Purified
GPNMB
Purified recombinant fragment of human GPNMB (AA: 31-260) expressed in E. Coli.
Protein A or G purified
Cell Cycle and Replication
Primary
10457
ELISA, Immunohistochemistry, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
glycoprotein (transmembrane) nmb, glycoprotein NMB, HGFINglycoprotein nmb-like protein, NMBosteoactivin, Transmembrane glycoprotein HGFIN, transmembrane glycoprotein NMB
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human