Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Osteoactivin/GPNMB Rabbit anti-Human, Clone: 105, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | Osteoactivin/GPNMB |
---|---|
Clone | 105 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Osteoactivin/GPNMB Monoclonal specifically detects Osteoactivin/GPNMB in Human samples. It is validated for ELISA.Specifications
Osteoactivin/GPNMB | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
glycoprotein (transmembrane) nmb, glycoprotein NMB, HGFINglycoprotein nmb-like protein, NMBosteoactivin, Transmembrane glycoprotein HGFIN, transmembrane glycoprotein NMB | |
Recombinant Human Osteoactivin/GPNMB Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
105 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
0.2 um filtered solution in PBS | |
10457 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title