Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Osteoadherin/OSAD/OMD Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795875PLATES

 View more versions of this product

Catalog No. NB010214


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Osteoadherin/OSAD/OMD.
Specifications

Specifications

HRP
Antibody Pairs
12.5 pg/mL
4958
RUO
Osteoadherin/OSAD/OMD
5 Plates
12.5-800 pg/mL
Keratan sulfate proteoglycan osteomodulin, KSPG osteomodulin, OSAD, Osteoadherin, osteoadherin proteoglycan, osteomodulin, SLRR2Costeoadherin
Conjugated
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Osteoadherin/OSAD/OMD.
Sandwich ELISA
  • Detection Antibody: 0.5 mg/mL rabbit anti-Osteoadherin/OSAD/OMD monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-mouse Osteoadherin/OSAD/OMD monoclonal antibody.
  • Standard: Each vial contains 232 ng of recombinant mouse Osteoadherin/OSAD/OMD.
  • Storage is content dependent.
    Rabbit
    Mouse
    OMD
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.8 ng/mL is recommended.
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit