Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Osteocalcin (Glu17) Mouse anti-Human, Unconjugated, Clone: 4B6, MilliporeSigma™
Mouse Monoclonal Antibody
$184.00 - $463.00
Specifications
Antigen | Osteocalcin (Glu17) |
---|---|
Clone | 4B6 |
Applications | ELISA, Dot Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS19672MI
|
MilliporeSigma
MABS196725UL |
25 μL |
Each for $184.00
|
|
|||||
MABS19671MI
|
MilliporeSigma
MABS1967100UL |
100 μL |
Each for $463.00
|
|
|||||
Description
Osteocalcin (Glu17) Monoclonal antibody specifically detects Osteocalcin (Glu17) in Human samples. It is validated for ELISA, Dot Blot.Specifications
Osteocalcin (Glu17) | |
ELISA, Dot Blot | |
Unconjugated | |
Mouse | |
Human | |
Bone Gla protein;BGP;Gamma-carboxyglutamic acid-containing protein | |
A linear peptide corresponding to 17 amino acids from the central region of the mature human osteocalcin with decarboxylated GLU17 residue (with signal peptide and pro regions cleaved off). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
10.96 kDa calculated (for complete molecule) |
4B6 | |
Monoclonal | |
Purified | |
Signaling | |
NP_954642.1 | |
BGLAP | |
IgG1 κ | |
Protein G Purified | |
Clone 4B6 is a mouse monoclonal antibody that detects human osteocalcin only when not carboxylated on the first glutamic acid residue (GLU17). It does not react with carboxylated osteocalcin. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title