Learn More
Enzo Life SciencesSupplier Diversity Partner Osteopontin (rodent), ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900090A
The Osteopontin (rodent), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Alternative Name: OPN. SPP1. Secreted phosphoprotein 1. Bone sialoprotein 1. Sensitivity: 3.03 ng/ml (range 3.13 - 100 ng/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Mouse, Rat. Contents: Microtiter plate, Standard, Antibody, Conjugate, Assay buffer 35, TMB Substrate, Stop solution 2, Wash buffer concentrate
Catalog No. 50-658-135