Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osterix/Sp7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen Osterix/Sp7
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126760
View Documents
Novus Biologicals
NBP31093025UL
25 μg
Each for $106.19
Add to cart
 
NB126759
View Documents
Novus Biologicals
NBP310930100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

Osterix/Sp7 Polyclonal antibody specifically detects Osterix/Sp7 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Osterix/Sp7
Western Blot, Immunohistochemistry
Purified
RUO
PBS buffer, 2% sucrose
121340
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Human
MGC126598, OSXosterix, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix
The immunogen is a synthetic peptide directed towards the N terminal region of human Osterix/Sp7 (NP_690599). Peptide sequence MASSLLEEEVHYGSSPLAMLTAACSKFGGSSPLRDSTTLGKAGTKKPYSV
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit