Learn More
OTC Mouse anti-Human, Clone: OTI1F3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802680
Catalog No. 50-166-6097
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.
Specifications
Specifications
OTC
Monoclonal
Unconjugated
OTC
Lyophilized
OTC
Human recombit protein fragment corresponding to amino acids 33-354 of human OTC produced in SF9 cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1F3
PBS with 8% trehalose and no preservative; pH 7.3
P00480
OCTD
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5009