Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTOA Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548131

Catalog No. PA548131

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May act as an adhesion molecule.
Specifications

Specifications

OTOA
Polyclonal
PBS with 0.09% sodium azide
Q7RTW8, Q8K561
Otoancorin, OTOA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
146183, 246190
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
OTOA
Liquid
OTOA
KLH conjugated synthetic peptide between 92-120 amino acids from the N-terminal region of human OTOA.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit