Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Otoferlin Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.03 - $436.00
Specifications
Antigen | Otoferlin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1: 300 - 1: 2000, Immunohistochemistry-Paraffin 1 : 300- 1 : 2000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
Otoferlin Polyclonal specifically detects Otoferlin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Otoferlin | |
Western Blot 1:300-1:2000, Immunohistochemistry 1: 300 - 1: 2000, Immunohistochemistry-Paraffin 1 : 300- 1 : 2000 | |
Polyclonal | |
Antisera | |
RUO | |
Whole antisera with No Preservative | |
9381 | |
A synthetic peptide from n-terminal cytoplasmic region of human Otoferlin (FER1L2) conjugated to an immunogenic carrier protein was used as the antigen. | |
Primary | |
Reconstitute with 0.1 ml sterilized water to desired concentration. | |
Specific for Otoferlin. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
AUNB1, DFNB6, DFNB9, EC 2.7.10.1, EC 4.2.1.11, FER1L2NSRD9, Fer-1-like protein 2, otoferlin | |
OTOF | |
IgG | |
Unpurified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title