Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTOP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA562727

Catalog No. PIPA562727


Add to cart

Description

Description

Immunogen sequence: DESVHQSHSY SSSHSNASHA RLISDQHADN PVGGDSCLCS TAVCQIFQQ Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 60%.

Specifications

Specifications

OTOP2
Polyclonal
Unconjugated
OTOP2
Liquid
OTOP2
Recombit protein corresponding to Human OTOP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q7RTS6
Otopetrin-2, OTOP2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
92736
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only