Learn More
OTOP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 188191AP
Catalog No. 50-173-2239
Add to cart
Description
Description
OTOP3 belongs to the otopetrin family.
Specifications
Specifications
OTOP3
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7RTS5, Q80UF9
OTOP3, otopetrin 3
Rabbit
IgG
150 μL
-20°C
Primary
347741, 69602
Western Blot
Unconjugated
OTOP3
Liquid
OTOP3
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse