Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUB2 Mouse anti-Canine, Human, Rat, Clone: OTI11B3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501946S

Catalog No. 50-167-3042


Add to cart

Description

Description

Hydrolase that can remove conjugated ubiquitin from proteins in vitro and may therefore play an important regulatory role at the level of protein turnover by preventing degradation. Mediates deubiquitination of  ft.Lys-11 ft.-, ft.Lys-48 ft.- and  ft.Lys-63 ft.-linked polyubiquitin chains, with a preference for  ft.Lys-63 ft.-linked polyubiquitin chains.
Specifications

Specifications

OTUB2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96DC9
C14orf137, OTB2, OTU2
Mouse
IgG1
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
314405, 480420, 78990
Flow Cytometry, Western Blot
OTI11B3
Unconjugated
OTUB2
Liquid
Otub2
Full length human recombit protein of human OTUB2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit