Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OTUB2 Mouse anti-Canine, Human, Rat, Clone: OTI11B3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA501946
Description
Hydrolase that can remove conjugated ubiquitin from proteins in vitro and may therefore play an important regulatory role at the level of protein turnover by preventing degradation. Mediates deubiquitination of ′Lys-11′-,′Lys-48′- and ′Lys-63′-linked polyubiquitin chains, with a preference for ′Lys-63′-linked polyubiquitin chains.Specifications
OTUB2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96DC9 | |
OTUB2 | |
Full length human recombit protein of human OTUB2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI11B3 | |
Unconjugated | |
OTUB2 | |
C14orf137, OTB2, OTU2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
314405, 480420, 78990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction