Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OTUD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525888
Description
OTUD4 Polyclonal Antibody for Western Blot, IHC (P)
OTUD4 alternatively spliced transcript variants have been found for this gene. The smaller protein isoform encoded by the shorter transcript variant is found only in HIV-1 infected cells.
Specifications
OTUD4 | |
Polyclonal | |
Unconjugated | |
OTUD4 | |
4930431L18Rik; AI449692; D8Ertd69e; DUBA6; HIN1; HIN-1; HIV-1 induced protein HIN-1; HIV-1-induced protein HIN-1; HSHIN1; KIAA1046; mKIAA1046; OTU deubiquitinase 4; OTU domain containing 4; OTU domain-containing protein 4; Otud4; RGD1305606 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
54726 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q01804 | |
OTUD4 | |
KLH conjugated synthetic peptide between 403-430 amino acids from the N-terminal region of human OTUD4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction