Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUD7A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548973

Catalog No. PA548973

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Has deubiquitinating activity towards  ft.Lys-11 ft.-linked polyubiquitin chains.
Specifications

Specifications

OTUD7A
Polyclonal
PBS with 0.09% sodium azide
Q8TE49
OTU domain-containing protein 7A, Zinc finger protein Cezanne 2, OTUD7A, C15orf16, CEZANNE2, OTUD7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
161725
Western Blot
Unconjugated
OTUD7A
Liquid
OTUD7A
KLH conjugated synthetic peptide between 70-99 amino acids from the N-terminal region of human OTUD7A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit