Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTX1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548716

Catalog No. PA548716

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probably plays a role in the development of the brain and the sense organs. Can bind to the BCD target sequence (BTS): 5 ft.-TCTAATCCC-3 ft..
Specifications

Specifications

OTX1
Polyclonal
PBS with 0.09% sodium azide
P32242
Homeobox protein OTX1, Orthodenticle homolog 1, OTX1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5013
Western Blot
Unconjugated
OTX1
Liquid
OTX1
KLH conjugated synthetic peptide between 87-116 amino acids from the Central region of human OTX1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit