Learn More
OTX2 Mouse anti-Human, Clone: OTI2B3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809510
Catalog No. 50-166-9546
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
OTX2 is a 289 amino acid containing protein encoding a member of the bicoid sub-family of homeodomain-containing transcription factors. It has two known alternative splice variants with distinct isoforms though presence of other isoforms cannot be excluded. The protein is an early marker of endodermal differentiation of embryonic stem cells and may play a role in the development of the brain and the sensory organs. The proteinmay bind to BCD target sequence (BTS): 5- ft.TCTAATCCC-3 ft.. Defects in OTX2 are the cause of microphthalmia syndromic type 5 (MCOPS5).
Specifications
Specifications
OTX2
Monoclonal
Unconjugated
OTX2
Lyophilized
OTX2
Full length human recombit protein of human OTX2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI2B3
PBS with 8% trehalose and no preservative; pH 7.3
P32243
CPHD6, MCOPS5
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5015