Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Otx2 Mouse anti-Human, DyLight 755, Clone: PCRP-OTX2-1E10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314028IR
This item is not returnable.
View return policy
Description
Otx2 Monoclonal antibody specifically detects Otx2 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Otx2 | |
Monoclonal | |
DyLight 755 | |
homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2 | |
Recombinant human full-lengthOtx2protein (Uniprot: P32243) | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers, Vision | |
5015 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-OTX2-1E10 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title