Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OTX3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$208.00 - $487.50
Specifications
| Antigen | OTX3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18025520
![]() |
Novus Biologicals
NBP18025520UL |
20 μL |
Each for $208.00
|
|
|||||
NBP180255
![]() |
Novus Biologicals
NBP180255 |
100 μL |
Each for $487.50
|
|
|||||
Description
OTX3 Polyclonal specifically detects OTX3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| OTX3 | |
| Polyclonal | |
| Rabbit | |
| NP_001020738 | |
| 127343 | |
| Synthetic peptide directed towards the middle region of mouse DMBX1. Peptide sequence LSLSEQSASESAPEDQLDREEDSRAEEPKAEKSPGSESKVPGCKRGSPKA. | |
| Primary |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| diencephalon/mesencephalon homeobox 1, diencephalon/mesencephalon homeobox protein 1, homeoprotein MBX, MBX, Orthodenticle homolog 3, OTX3, Paired-like homeobox protein DMBX1, PAXBorthodenticle homolog 3 (Drosophila) | |
| DMBX1 | |
| IgG | |
| 41 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title