Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Ouchterlony Plates
Designed for specific but low affinity antisera that requires properly spaced wells and olyethylene glycol in the agarose for optimal immuno -precipitation.
$396.00
Specifications
Material | Polystyrene |
---|---|
Type | Ouchterlony Plate |
Includes | Agarose 1.2% and PEG 5% |
Catalog Number | Mfr. No. | For Use With (Application) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Type | Price | Quantity & Availability | ||||
ICN642771
|
MP Biomedicals™
08642771 |
Specific and low affinity antisera | Ouchterlony Plate |
Each of 1 for $396.00
|
|
Specifications
Polystyrene | |
Ouchterlony Plate | |
Agarose 1.2% and PEG 5% |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title