Learn More
ILC Dover OV/AG Cartridge Filters, Set of 3
The OV/AG Cartridge Fitlers protect against Particulates, Organic Vapors, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, and Hydrogen Fluoride.
Supplier:  ILC Dover S4012
Catalog No. 19-631-085
Only null left
Add to Cart
Description
Description
  • Protect against Particulates, Organic Vapors, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, and Hydrogen Fluoride
Specifications
Specifications
Combination Cartridge/Filter
3/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.