Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ovalbumin Mouse anti-Chicken, Alexa Fluor™ 350, Clone: 8C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223533AF350

Catalog No. NP22333A30


Add to cart

Description

Description

Ovalbumin Monoclonal antibody specifically detects Ovalbumin in Chicken samples. It is validated for ELISA, Western Blot.
Specifications

Specifications

Ovalbumin
8C6
ELISA, Western Blot (Negative)
Conjugated, Purified
OVAL
Purified chicken ovalbumin adsorbed onto aluminum hydroxide gel
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Clone 8C6 reacts with native chicken ovalbumin. Cross-reactivity with ovalbumins from other avian species has not been tested
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8
Mouse
IgG1 Kappa
0.1 mL
Primary
396058.0
Chicken
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit