Learn More
US Biomax Inc OVARY CANCER WITH OVARY TISSUE  DFS Item
Manufacturer:  US Biomax Inc OV809B
Ovary cancer with ovary tissue array, including pathology grade, TNM and clinical stage, 80 cases/80 cores, replacing OV809a
Catalog No. NC1734873