Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Oven Shroud

Includes Heating Coil and Oven Temperature Sensor
Supplier: Restek™ 27187
Description
- Available in 120 V and 220 V units
- For Agilent 6890/7890 GCs
Specifications
220 V | |
Heating Coil and Oven Temperature Sensor | |
Agilent 6890/7890 GCs |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title