Learn More
BioworldSupplier Diversity Partner OVINE LEPTIN TRIPLE ANTAGON DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220606121
OVINE LEPTIN TRIPLE ANTAGONIST (RECOMBINANT): Recombinant ovine leptin (oLEP) antagonist (triple mutant) Source: E.Coli Background: Leptin inhibits food intake and stimulates energy expenditure. Leptin also has thermogenic actions an...
Catalog No. 50-253-712