Learn More
Raybiotech IncSupplier Diversity Partner Ovine VEGF-A ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELOVEGFA1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Ovine VEGF-A.
Catalog No. 50-195-0176