Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OVOS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571560
Description
OVOS2 Polyclonal Antibody for Western Blot, IHC (P)
Is able to inhibit all four classes of proteinases by a unique 'trapping' mechanism. [UniProt].
Specifications
OVOS2 | |
Polyclonal | |
Unconjugated | |
OVOS2 | |
OVOS2; ovostatin 2; ovostatin homolog 2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
144203 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q6IE36 | |
OVOS2 | |
KLH conjugated synthetic peptide between 99-128 amino acids from the N-terminal region of human OVOS. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction