Thermo Scientific™ Owl™ A1 Large Gel System Available on GSA/VA Contract for Federal Government customers only.

Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems, which have smaller footprints ideal for detailed RNA/DNA analysis.

Manufacturer: Thermo Scientific™ A1

 View more versions of this product

Catalog No. 09-528-100

  Check Availability
Add to Cart

    Specifications

    Description Complete system
    Height (Metric) Exterior 11cm
    Length (English) Exterior 14.8 in.
    Width (English) Exterior 8.5 in.
    Length (Metric) Gel 25cm
    Width (Metric) Gel 13cm
    Height (English) Exterior 4.3 in.
    Dimensions (L x W x H) Exterior 14.8 x 8.5 x 4.3 in. (37.5 x 21.5 x 11cm)
    Length (Metric) Exterior 37.5cm
    Width (Metric) Exterior 21.5cm
    Certifications/Compliance CE marked
    Type Large
    Sample Range 8 to 150
    Volume (Metric) Buffer 1.6L

    Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems. With one of the smallest footprints of any large-format device, these systems are ideal for detailed RNA/DNA analysis.

    Features:

    • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
    • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
    • Programmable power inverter may be used for field reversal electrophoresis
    • UVT gel tray has gasketed end gates for leakproof seal without tape
    • Built-in leveling ensures flat, uniformly thick gels
    • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
    • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
    • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time

    Includes:

    • Buffer chamber with three-point leveling base
    • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
    • UVT gel tray
    • Gasketed end gates
    • Three combs (12-, 16- and 20-well), 1.5mm thick

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit