Learn More
Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts Available on GSA/VA Contract for Federal Government customers only.
Choose from various Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts, including gasketed UVT gel trays, replacement gaskets and external casters.
$28.85 - $244.50
Specifications
For Use With (Equipment) Owl™ A1 Large Gel Electrophoresis System
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
OW-A1GK Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
A1GK
Replacement Gaskets (1 pair)
Pack of 2 for $28.85
Add to cart
OW-A1UVT Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
A1UVT
UVT tray with gasketed end gates
Each for $244.50
Add to cart
Description
Description
Choose from various Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts, including gasketed UVT gel trays, replacement gaskets and external casters.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned so you can arrange up to 10 sample sets on the same gel
  • Unique electrode placement allows this gel to be run quickly without compromising the resolution
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Owl™ A1 Large Gel Electrophoresis System