Learn More
Thermo Scientific™ Owl™ A2-OK Multiple Gel Electrophoresis System
Run multiple gels simultaneously with Thermo Scientific™ Owl™ A2-OK Multiple Gel Electrophoresis Systems, which are also ideal for teaching labs.
Supplier: Thermo Scientific™ A2OK
Description
Run multiple gels simultaneously with Thermo Scientific™ Owl™ A2-OK Multiple Gel Electrophoresis Systems, which are also ideal for teaching labs.
Key Features:- Up to six 7 x 8cm minigels can be run on one system
- Screen up to 120 samples at once with supplied combs
- Thermo Scientific™ EasyCast™ casting design permits casting without tape or plugs
- UV-transmissible (UVT) gel trays may be placed directly on the transilluminator for easy photodocumentation
- Teacher can instruct up to 12 students using one system and one power supply
Includes:
Buffer chamber with three-point leveling base;Thermo Scientific™ SuperSafe™ lid with attached power supply leads; Six EasyCast UVT gasketed gel trays; Twelve 10-well, double-sided combs, 1.0/1.5mm thick; Two external multiple gel casters; Gel tray holder
Specifications
CE marked | |
7 cm x 8 cm | |
Multiple Gel Electrophoresis System | |
2.3 L | |
14.8 in. | |
8 cm | |
32.5 cm | |
4.1 in. |
Gel Size: Up to six 7 x 8cm; Buffer volume: 2.3L; L x W x H: 37.5 × 32.5 × 10.5cm | |
Owl™ | |
5 to 144 | |
14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm) | |
37.5 cm | |
12.8 in. | |
7 cm | |
10.5 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.