Learn More
Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts
Choose these replacement parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
$31.95 - $408.50
Specifications
For Use With (Equipment) Owl™ A3-1 and A6 Wide Gel System
View More Specs
Products 3
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
09-528-204
View Documents
Thermo Scientific™
A31EG
Replacement end gates (1 pair)
Pack of 2 for $101.80
Add to cart
OW-A3-GK-1
View Documents
Thermo Scientific™
A3GK1
Replacement Gaskets (1 pair)
Pack of 2 for $31.95
Add to cart
OW-A3UVT1
View Documents
Thermo Scientific™
A3UVT1
UVT gel tray with gasketed end gates
Each for $408.50
Add to cart
Description
Description
Choose Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
The gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run utilizing a wall comb. Built in buffer exchange ports allows buffer recirculation on extended runs, if needed. The large number of comb slots gives you sample and resolving distance flexibility.
Specifications
Specifications
Owl™ A3-1 and A6 Wide Gel System