Learn More
Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Combs
Eliminate comb assembly with these heavy-duty, one-piece, standard seven-tooth combs.
$83.50
Specifications
Description Standard comb
For Use With (Equipment) Owl™ C2-S Micro Electrophoresis System
Teeth Count 7
View More Specs
Products 2
Catalog Number Mfr. No. Description Teeth Count Thickness (Metric) Teeth Price Quantity & Availability  
Catalog Number Mfr. No. Description Teeth Count Thickness (Metric) Teeth Price Quantity & Availability  
OW-C27C
View Documents
Thermo Scientific™
C27C
Standard comb 7 1.0 mm
Each for $83.50
Add to cart
OW-C27D
View Documents
Thermo Scientific™
C27D
Standard comb 7 1.5 mm
Each for $83.50
Add to cart
Description
Description
Eliminate comb assembly with heavy-duty, one-piece Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Combs. These standard seven-tooth combs are available in two thicknesses (1.0 and 1.5mm) and are for use with the Owl™ C2-S Micro Electrophoresis System.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifications
Specifications
Standard comb
Owl™ C2-S Micro Electrophoresis System
7