Learn More
Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts Available on GSA/VA Contract for Federal Government customers only.
Replace casting slides, casting tray or buffer chamber with Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts.
$37.40 - $105.70
Specifications
For Use With (Equipment) Owl™ C2-S Micro Electrophoresis System
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
OW-C2CST
View Documents
Thermo Scientific™
C2CST
Gel casting tray Each for $105.70
Add to cart
09-528-125 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
CGS
Casting slides; Pack of 10 Pack of 10 for $37.40
Add to cart
Description
Description
Replace casting slides, casting tray or buffer chamber with Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts. Parts are for use with the Owl C2-S Micro Electrophoresis System, a system designed for quick restriction fragment analysis.
  • Glass slides, 5 x 7.5 cm, are used as the gel tray
  • Thin gels can be poured and formed by surface tension directly on the glass slide
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Owl™ C2-S Micro Electrophoresis System