Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs Available on GSA/VA Contract for Federal Government customers only.
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Manufacturer:  Thermo Scientific™ D130C
 View more versions of this product
Catalog No. 09-528-167F
Add to cart
Description
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Micro well
Micro well; 30 (2X) teeth; 1.0mm thick
1mm
Combs
Thermo Scientific™ Owl D2 Wide-Gel Electrophoresis Systems
30 (2X)
Owl™ D2 Wide-Gel Electrophoresis Systems