Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Manufacturer: Thermo Scientific™ D120D
 View more versions of this product
Catalog No. 09-528-167C
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Owl™ D2 Wide-Gel Electrophoresis Systems
    Type Combs
    Description Standard
    For Use With Thermo Scientific™ Owl D2 Wide-Gel Electrophoresis Systems
    Item Description Standard; 20 teeth; 1.5mm thick
    Teeth Count 20
    Thickness (Metric) Teeth 1.5mm
    Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
    Heavy-duty, one-piece combs eliminate comb assembly.
  • Description & Specifications
Related Products
Product Content Feedback