Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty one-piece combs, for use with the Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System.
Manufacturer: Thermo Scientific™ B112D
 View more versions of this product
Catalog No. 09-528-180
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Owl™ EasyCast B1 Mini Gel Electrophoresis System
    Type Combs
    Description Standard comb
    For Use With Thermo Scientific™ Owl EasyCast B1 Mini Gel Electrophoresis System
    Item Description Standard 12 tooth comb; 1.5mm thick
    Teeth Count 12
    Thickness (Metric) Teeth 1.5mm
    Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs. These heavy-duty one-piece combs are for use with the EasyCast™ B1 Mini Gel Electrophoresis System.
    Heavy-duty, one-piece combs eliminate comb assembly.
    Ordering Information:
    Cat Nos. B1-Rl-9C, -9D, -17C and -17D are 8 and 12-channel pipette format (1X) and (2X) combs.
    Customized combs are availabe (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
  • Description & Specifications
Related Products
Product Content Feedback