Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems Available on GSA/VA Contract for Federal Government customers only.
Owl EasyCast B2 Mini Gel Electrophoresis Systems provide superior flexibility in a small footprint.
Manufacturer:  Thermo Scientific™ B2
 View more versions of this product
Catalog No. 09-528-110B
Add to cart
Description
Description
Thermo Scientific™ Owl EasyCast B2 Mini Gel Electrophoresis Systems provide superior flexibility in a small footprint. The simple to use casting method, and multiple comb and tray options, makes the EasyCast B2 Owl's most popular system.
The EasyCast systems feature leakproof casting that allows a gel to be cast and run in the same chamber with no tape or additional parts required. The EasyCast B2's longer gel length and the 2 comb slots provide the flexibility of running 8 to 48 samples on 1 gel. The optional multi load tray can increase the maximum number of samples to 108 in an 8 or 12 multichannel pipette format.
  • 2 comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands
Ordering Information:
Available with optional buffer exchange ports (B2-BP) for use with an external recirculation pump
Includes:
Buffer chamber, SuperSafe™ lid with attached power supply leads, EasyCast gasketed U.V. Transmissible (UVT) gel tray, 2 combs: 12 & 20 well, double-sided, 1.0/1.5 mm thick
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
EasyCast
Mini
800mL
12cm
9.5cm
24.5cm
18cm
9.63 x 7.06 x 3.75 in. (24.5 x 18 x 9.5cm)
8 to 48
14cm
3.75 in.
9.63 in.
7.06 in.
CE marked
Documents
Accessories