Learn More
Thermo Scientific™ Owl™ Gel-Drying Kits
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
Manufacturer:  Thermo Scientific™ GDF10
 View more versions of this product
Catalog No. OW-GDF-10
Add to cart
Description
Description
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures. Thermo Scientific™ Owl™ Gel-Drying Kits provide an inexpensive and simple way to produce a permanent record of gels by air drying them between two sheets of cellophane.
  • Available in minigel and large format sizes
  • Convenient method of preserving gels by air drying them between two sheets of cellophane
  • Beveled-edge design of gel-drying frame makes assembly easy
Includes:
Complete kits include two sets of gel-drying frames, 50 sheets of precut cellophane, two solid back plates, and eight side clips; frames and cellphane are also available separately
Specifications
Specifications
Gel-Drying Kit, Minigel Format
14 cm
Gel Drying Kit
14 x 14 cm
14 cm
Each