Learn More
Thermo Scientific™ Owl™ Gel-Drying Kits Available on GSA/VA Contract for Federal Government customers only.
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
Manufacturer:  Thermo Scientific™ GDF20
 View more versions of this product
Catalog No. OW-GDF-20
Add to cart
Description
Description
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures. Thermo Scientific™ Owl™ Gel-Drying Kits provide an inexpensive and simple way to produce a permanent record of gels by air drying them between two sheets of cellophane.
  • Available in minigel and large format sizes
  • Convenient method of preserving gels by air drying them between two sheets of cellophane
  • Beveled-edge design of gel-drying frame makes assembly easy
Includes:
Complete kits include two sets of gel-drying frames, 50 sheets of precut cellophane, two solid back plates, and eight side clips; frames and cellphane are also available separately
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Gel-Drying Kit, Large Format
24 cm
Gel Drying Kit
24 x 24 cm
24 cm
Each