Learn More
Thermo Scientific™ Owl™ Gel-Drying Kits
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
$196.40 - $280.00
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
OW-GDF-10
View Documents Promotion Details
Thermo Scientific™
GDF10
Gel-Drying Kit, Minigel Format
Each for $196.40
Add to cart
OW-GDF-20
View Documents Promotion Details
Thermo Scientific™
GDF20
Gel-Drying Kit, Large Format
Each for $280.00
Add to cart
Description
Description
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures. Thermo Scientific™ Owl™ Gel-Drying Kits provide an inexpensive and simple way to produce a permanent record of gels by air drying them between two sheets of cellophane.
Features:
  • Available in minigel and large format sizes
  • Beveled-edge design of gel-drying frame makes assembly easy
Includes:
  • Two sets of gel-drying frames
  • 50 sheets of precut cellophane
  • Two solid back plates
  • Eight side clips
Note:
  • Frames and cellophane are available separately