Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Owl™ Gel-Drying Supplies
Use the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.
Supplier: Thermo Scientific™ CM10
Description
Combine the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.
- Frames and pre-cut cellophane available in large (24 × 24cm) or small (14 × 14cm) format
Specifications
Precut Cellophane; 14 x 14cm | |
Cellophane, Precut | |
14 cm |
14 x 14 cm | |
14 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction