Learn More
Thermo Scientific™ Owl™ S3S System Combs
Eliminate comb assembly with these heavy-duty, one-piece combs.
$122.90
Specifications
For Use With (Equipment) | Owl™ S3S Aluminum-Backed Sequencer System |
---|---|
Product Type | Combs |
Catalog Number | Mfr. No. | Description | Teeth Count | Thickness (Metric) Teeth | Volume (Metric) Well | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Teeth Count | Thickness (Metric) Teeth | Volume (Metric) Well | Price | Quantity & Availability | ||
OW-S2S-MT2
|
Thermo Scientific™
S2SMT2 |
Sharktooth Micro-Well Comb | 72 (2X) | 0.4 mm | 10µL |
Each for $122.90
|
|
OW-S2S-MT3
|
Thermo Scientific™
S2SMT3 |
Sharktooth Comb | 110 (3X) | 0.4 mm | 5.5µL |
Each for $122.90
|
|
OW-S2S-MT44
|
Thermo Scientific™
S2SMT44 |
Sharktooth Micro-Well Comb | 146 (4X) | 0.4 mm | 3.7µL |
Each for $122.90
|
|
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ S3S System Combs. These heavy-duty, one-piece combs are for use with the Owl™ S3S System.
Heavy duty, one piece combs eliminate comb assembly.Specifications
Owl™ S3S Aluminum-Backed Sequencer System | |
Combs |