Learn More
OX40 Ligand/TNFSF4, Human anti-Human, PE-Atto594, Clone: R4930 (Oxelumab), Novus Biologicals™
Human Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP275913PEATT594100UL
Catalog No. NB009332
Add to cart
Description
Description
OX40 Ligand/TNFSF4 Monoclonal antibody specifically detects OX40 Ligand/TNFSF4 in Human Samples. It is validated for Flow Cytometry.
Specifications
Specifications
OX40 Ligand/TNFSF4
R4930 (Oxelumab)
Flow Cytometry
Purified
TNFSF4
Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells.
Protein A purified
RUO
Store at 4°C in the dark. Do not freeze.
Monoclonal
Binds to human OX40L.
Flow Cytometry
PE/ATTO 594
PBS
CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Human
IgG1 Kappa
100 uL
Cancer, Cell Cycle and Replication, Immunology
Primary
7292
Human