Learn More
OX40 Ligand/TNFSF4 Rabbit anti-Human, Alexa Fluor™ 532, Clone: R4930 (Oxelumab), Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252687AF532
Catalog No. NP22687A32
Add to cart
Description
Description
OX40 Ligand/TNFSF4 Monoclonal antibody specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
OX40 Ligand/TNFSF4
R4930 (Oxelumab)
Flow Cytometry
CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Binds to human OX40L.
Flow Cytometry
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
TNFSF4
Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells.
0.1 mL
Primary
7292
Human