Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OX40 Ligand/TNFSF4 Rabbit anti-Human, Alexa Fluor™ 750, Clone: R4930 (Oxelumab), Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252687AF750

Catalog No. NP22687A70


Add to cart

Description

Description

OX40 Ligand/TNFSF4 Monoclonal antibody specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

OX40 Ligand/TNFSF4
R4930 (Oxelumab)
Flow Cytometry
Conjugated, Purified
TNFSF4
Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells.
Protein A or G purified
Cancer, Cell Cycle and Replication, Immunology
Primary
7292.0
Human
Flow Cytometry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Rabbit
IgG Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Binds to human OX40L.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit